Thermo Scientific Chemicals

Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: 044410.30
250 g, Each
Thermo Scientific Chemicals

Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: 044410.30
250 g, Each
Quantity
Catalog number: 044410.30
also known as 044410-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144-62-7
IUPAC Name
oxalic acid
Molecular Formula
C2H2O4
InChI Key
MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES
OC(=O)C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

Oxalic acid is used as reducing agent for photographic films, as mordant in dyeing process, as bleaching agent for removal of rust through complex formation, and as grinding agent during polishing of marble. It is used in waste water treatment to remove calcium. The conjugate base of oxalic acid acts as a chelating agent for metal ions. It is used as a catalyst for esterifications in synthetic organic chemistry and as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text