Thermo Scientific Chemicals

Europium(II) bromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044431.06
5 g, Each
Thermo Scientific Chemicals

Europium(II) bromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044431.06
5 g, Each
Quantity
Catalog number: 044431.06
also known as 044431-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13780-48-8
IUPAC Name
λ²-europium(2+) dibromide
Molecular Formula
Br2Eu
InChI Key
PJVPGMOCWCUQHP-UHFFFAOYSA-L
SMILES
[Br-].[Br-].[Eu++]
Total Metal Impurities
0.01% max.

Description

Europium(II) bromide is used in water treatment, chemical analysis and in certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text