Thermo Scientific Chemicals

Cerium(III) bromide hydrate, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 044432.18
50 g, Each
Thermo Scientific Chemicals

Cerium(III) bromide hydrate, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 044432.18
50 g, Each
Quantity
Catalog number: 044432.18
also known as 044432-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
396654-07-2
Total Metal Impurities
0.002% max.

Description

Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text