Thermo Scientific Chemicals

Cerium(III) acetate hydrate, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 044433.30
250 g, Each
Thermo Scientific Chemicals

Cerium(III) acetate hydrate, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 044433.30
250 g, Each
Quantity
Catalog number: 044433.30
also known as 044433-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
206996-60-3
IUPAC Name
cerium(3+) triacetate
Molecular Formula
C6H9CeO6
InChI Key
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
SMILES
[Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.005% max.

Description

Cerium(III) acetate hydrate is used in thin fim buffer layers suitable for high κ dielectrics by sol-gel route. And it is also used as a catalyst for auto-oxidation of cresols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text