Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals
Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 044434.14
also known as 044434-14
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
Add to cart
Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number044434.14
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3396-11-0
IUPAC Namecaesium(1+) acetate
Molecular FormulaC2H3CsO2
InChI KeyZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES[Cs+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Cesium acetate is used in organic synthesis especially in the Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only