Thermo Scientific Chemicals

Aluminum fluoride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044435.18
50 g, Each
Thermo Scientific Chemicals

Aluminum fluoride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044435.18
50 g, Each
Quantity
Catalog number: 044435.18
also known as 044435-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-18-1
IUPAC Name
aluminium(3+) trifluoride
Molecular Formula
AlF3
InChI Key
KLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Al+3]
Form
Crystals or powder or crystalline powder
Total Metal Impurities
≤0.01% (≤100ppm)
Elemental Analysis
Aluminum, Al: Reported (Theory 32.13%)
Appearance (Color)
White
Assay (special)
≤99.99% (metals basis)

Description

Aluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text