Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044436.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7719-12-2
IUPAC Nametrichlorophosphane
Molecular FormulaCl3P
InChI KeyFAIAAWCVCHQXDN-UHFFFAOYSA-N
SMILESClP(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
Phosphorus(III) chloride is used as a reagent in organic synthesis. It is utilized in the conversion of primary alcohol and secondary alcohol to alkyl chlorides. It acts as a precursor to phosphorous pentachloride, phosphorous oxychloride, thiophosphoryl chloride and triphenylphosphine. It is also used to prepare phosphite esters, which is used Horner-Wadsworth-Emmons reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only