Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044442.22
100 g, Each
Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044442.22
100 g, Each
Quantity
Catalog number: 044442.22
also known as 044442-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13446-34-9
IUPAC Name
manganese(2+) tetrahydrate dichloride
Molecular Formula
Cl2H8MnO4
InChI Key
CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES
O.O.O.O.[Cl-].[Cl-].[Mn++]
Total Metal Impurities
0.01% max.

Description

It is used as a source of manganese in biological research, in the production of dry cell batteries, and precursor to the antiknock compound. It is used in industrial applications such as dyeing disinfecting, the purification of natural gas, and as a drying agent for linseed oil. It is used in pigment manufacturing, fertilizers and in construction materials and also used in aluminum smelting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text