Thermo Scientific Chemicals

3-Nitrobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 044445.06
5 g, Each
Thermo Scientific Chemicals

3-Nitrobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 044445.06
5 g, Each
Quantity
Catalog number: 044445.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
619-25-0
IUPAC Name
(3-nitrophenyl)methanol
Molecular Formula
C7H7NO3
InChI Key
CWNPOQFCIIFQDM-UHFFFAOYSA-N
SMILES
OCC1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
Pale yellow to yellow to orange to pale brown
Assay from Suppliers CofA
≥98.5%
Form
Liquid or fused solid

Description

3-Nitrobenzyl alcohol is the best substrate for cytosolic alcohol dehydrogenase. 3-Nitrobenzyl alcohol was employed as matrix during fast-atom bombardment mass spectrometry. It was used in isolation of palmitylated peptide fragment from bovine rhodopsin and its characterization by mass spectrometry. It has been used as a liquid matrix for fast atom bombardment[1] and matrix-assisted laser desorption ionization. In electrospray ionization 3-NBA is doped into low surface tension spray solvents to increase analyte charging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text