Thermo Scientific Chemicals
4-Methylbenzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.0 mmol/g on PS-DVB, Thermo Scientific Chemicals
Catalog number: 044455.06
5 g, Each
Thermo Scientific Chemicals
4-Methylbenzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.0 mmol/g on PS-DVB, Thermo Scientific Chemicals
Catalog number: 044455.06
5 g, Each
Quantity
Catalog number: 044455.06
also known as 044455-06
Price (USD)
Quantity
-
Have Questions?
Description
Hydrochloride resins have been used to study the treatment of hyperphosphataemia in patients with end-stage renal failure. And also used for attachment of linkers in a similar manner to PL-AMS Resin.