Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044456.30
250 g, Each
Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044456.30
250 g, Each
Quantity
Catalog number: 044456.30
also known as 044456-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-69-4
IUPAC Name
lithium(1+) nitrate
Molecular Formula
LiNO3
InChI Key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES
[Li+].[O-][N+]([O-])=O
Form
Powder and/or granules
Assay (special)
?2.5 % of Theoretical value (Li)
Total Metal Impurities
≤0.001% (10 ppm)
Appearance (Color)
White

Description

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text