n-Heptane, anhydrous, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
n-Heptane, anhydrous, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Heptane, anhydrous, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 044457.AE
also known as 044457-AE
Price (USD)/ Each
40.65
Online exclusive
44.90 
Save 4.25 (9%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
44.90 
Save 4.25 (9%)
Add to cart
n-Heptane, anhydrous, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number044457.AE
Price (USD)/ Each
40.65
Online exclusive
44.90 
Save 4.25 (9%)
-
Add to cart
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥99%
CommentInert gas used can be either Argon or Nitrogen
Water Content (Karl Fischer Titration)≤10 ppm
Residue on Evaporation≤5 ppm
View more
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only