Thermo Scientific Chemicals

O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals

Catalog number: 044465.02
0.5 g, Each
Thermo Scientific Chemicals

O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals

Catalog number: 044465.02
0.5 g, Each
Quantity
Catalog number: 044465.02
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2511-10-6
IUPAC Name
ethyl (ethylsulfanyl)(methyl)phosphinate
Molecular Formula
C5H13O2PS
InChI Key
AIMOGGQLCCAPQJ-UHFFFAOYNA-N
SMILES
CCOP(C)(=O)SCC
Form
Liquid
Assay from Supplier's CofA
≥96.0%

Description

O,S-Diethyl methylphosphonothioate is used as an inhibitor of cholinesterase. It is also used in pharmaceuticals, intermediates and fine chemicals. It also serves as phosphorylating and phosphitylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text