Search Thermo Fisher Scientific
Cerium(IV) isopropoxide, Ce 37-45% is used to catalyzes diastereoselective pinacol couplings. It forms diketonates ,which hydrolyze to CeO2 particles. CeO2 particles have the potential for ion conduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.