Lithium hydroxide monohydrate, 99.995% (metals basis), Thermo Scientific Chemicals
Lithium hydroxide monohydrate, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, 99.995% (metals basis), Thermo Scientific Chemicals

Lithium hydroxide monohydrate, H3LiO2, CAS Number-1310-66-3, unii-g51xlp968g, lithium hydroxide monohydrate, lioh water, lithiumhydrate, hydroxyde de lithium, lithium hydroxido, lithium hydroxide, monohydrate, lithium hydroxide hydrate, lioh.hydrate, lioh-hydrate, 100g, 99.995% (Metals basis), White
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 044473.22
also known as 044473-22
Price (USD)/ Each
281.65
Online exclusive
313.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
281.65
Online exclusive
313.00 
Save 31.35 (10%)
Add to cart
Lithium hydroxide monohydrate, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number044473.22
Price (USD)/ Each
281.65
Online exclusive
313.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Trace Metals (by ICP)≤0.005% (≤50 ppm)
Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only