Thermo Scientific Chemicals

Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 98%, Thermo Scientific Chemicals

Catalog number: 044483.09
10 g, Each
Thermo Scientific Chemicals

Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 98%, Thermo Scientific Chemicals

Catalog number: 044483.09
10 g, Each
Quantity
Catalog number: 044483.09
also known as 044483-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80657-57-4
IUPAC Name
methyl 3-hydroxy-2-methylpropanoate
Molecular Formula
C5H10O3
InChI Key
ATCCIZURPPEVIZ-UHFFFAOYNA-N
SMILES
COC(=O)C(C)CO
Assay (GC)
≥ 97.5%
Optical Rotation
23.0 to 29.0?C ( c=4 in methanol)

Description

Methyl (S)-(+)-3-hydroxy-2-methylpropionate is used as a starting material for the synthesis of dictyostatin, discodermolide and spongidepsin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text