Hafnium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Hafnium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
250 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044489.DJ
Price (USD)/ Each
-
Quantity:
50 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialHafnium rod
Name Note6.35mm (0.25 in.) dia.
CAS7440-58-6
Recommended StorageAmbient temperatures
EINECS Number231-166-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only