Potassium silicate, anhydrous, Thermo Scientific Chemicals
Potassium silicate, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium silicate, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 044493.22
also known as 044493-22
Price (USD)/ Each
30.65
Online exclusive
33.50 
Save 2.85 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
33.50 
Save 2.85 (9%)
Add to cart
Potassium silicate, anhydrous, Thermo Scientific Chemicals
Catalog number044493.22
Price (USD)/ Each
30.65
Online exclusive
33.50 
Save 2.85 (9%)
-
Add to cart
Chemical Identifiers
CAS1312-76-1
IUPAC Namepotassium hydroxysilanoylolate
Molecular FormulaHKO3Si
InChI KeyWMWBFVSTSRDWRL-UHFFFAOYSA-N
SMILES[K+].O[Si]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)28.0-28.8% K2O
Potassium silicate is used in binders, carbon electrodes, lead pencils, protective coatings, insoluble pigments, phosphors on television tubes and detergent additives. It acts as a catalyst to make glass, ceramics, welding rods, adhesives, cements, gels, paints, pulp and paper production. Further, it is used in horticulture as a soluble source of potassium and silicon. It acts as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only