Platinum, 10% on alumina, reduced, Thermo Scientific Chemicals
Platinum, 10% on alumina, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum, 10% on alumina, reduced, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 044494.22
also known as 044494-22
Price (USD)/ Each
1,627.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
1,627.00
Add to cart
Platinum, 10% on alumina, reduced, Thermo Scientific Chemicals
Catalog number044494.22
Price (USD)/ Each
1,627.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPlatinum
Name Note10% on alumina, Reduced
CAS7440-6-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H341-H335
Suspected of causing genetic defects.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only