Calcium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Calcium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 044497.22
also known as 044497-22
Price (USD)/ Each
428.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
428.00
Add to cart
Calcium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number044497.22
Price (USD)/ Each
428.00
-
Add to cart
Chemical Identifiers
CAS71626-99-8
IUPAC Namecalcium dibromide
Molecular FormulaBr2Ca
InChI KeyWGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES[Ca++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only