Thermo Scientific Chemicals

mu-Chloro-mu-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, 0.5M in toluene, Thermo Scientific Chemicals

Catalog number: 044506.AC
25 mL, Each
Thermo Scientific Chemicals

mu-Chloro-mu-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, 0.5M in toluene, Thermo Scientific Chemicals

Catalog number: 044506.AC
25 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 044506.AC
also known as 044506-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67719-69-1
IUPAC Name
titanium(4+) (dimethylalumanyl)methanide bis(cyclopenta-2,4-dien-1-ide) chloride
Molecular Formula
C13H18AlClTi
InChI Key
HIVIAXIRSNAVAD-UHFFFAOYSA-M
SMILES
[Cl-].[Ti+4].C[Al](C)[CH2-].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Appearance (Color)
Red to dark red to purple
Form
Solution
Assay (unspecified)
0.40 to 0.60M

Description

μ-Chloro-μ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum is used in organic synthesis for carbonyl methylenation, A versatile methylenation reagent for the conversion of ketones and aldehydes to olefins, it offers facile reaction with hindered ketones and allows the conversion of esters to vinyl ethers. Tebbe reagent can be used to olefinate aldehydes, to methylenate a chiral polyhydroxyketone with high diasteroselctivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text