2-Hydroxymethyl-12-crown-4, 97%, Thermo Scientific Chemicals
2-Hydroxymethyl-12-crown-4, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxymethyl-12-crown-4, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
5 g
Catalog number 044509.03
also known as 044509-03
Price (USD)/ Each
111.65
Online exclusive
124.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
111.65
Online exclusive
124.00 
Save 12.35 (10%)
Add to cart
2-Hydroxymethyl-12-crown-4, 97%, Thermo Scientific Chemicals
Catalog number044509.03
Price (USD)/ Each
111.65
Online exclusive
124.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75507-26-5
IUPAC Name[(2S)-1,4,7,10-tetraoxacyclododecan-2-yl]methanol
Molecular FormulaC9H18O5
InChI KeyNJIPEIQHUNDGPY-VIFPVBQESA-N
SMILESOC[C@H]1COCCOCCOCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay from Supplier's CofA≥97% (GC)
FormLiquid
Ethylene oxide is important or critical to the production of detergents, thickeners, solvents, plastics, and various organic chemicals such as ethylene glycol, ethanolamines, simple and complex glycols, polyglycol ethers and other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only