Thermo Scientific Chemicals

Amidosulfonic acid, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044510.36
500 g, Each
Thermo Scientific Chemicals

Amidosulfonic acid, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044510.36
500 g, Each
Quantity
Catalog number: 044510.36
also known as 044510-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5329-14-6
IUPAC Name
sulfamic acid
Molecular Formula
H3NO3S
InChI Key
IIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILES
NS(O)(=O)=O
Form
Powder or crystals or crystalline powder
Total Metal Impurities
≤0.01%

Description

Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text