Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044515.14
also known as 044515-14
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
Add to cart
Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Catalog number044515.14
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS35054-52-5
IUPAC Namecalcium dinitrate
Molecular FormulaCaN2O6
InChI KeyZCCIPPOKBCJFDN-UHFFFAOYSA-N
SMILES[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0005% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only