Thermo Scientific Chemicals

Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion, Thermo Scientific Chemicals

Catalog number: 044517.A1
1 kg, Each
Thermo Scientific Chemicals

Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion, Thermo Scientific Chemicals

Catalog number: 044517.A1
1 kg, Each
Quantity
Catalog number: 044517.A1
also known as 044517-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13463-67-7
IUPAC Name
dioxotitanium
Molecular Formula
O2Ti
InChI Key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES
O=[Ti]=O
Form
Dispersion
Assay (special)
Solids: 20-36%

Description

UV protection in clear coatings. Photo electrodes of varying properties can be made using colloidal suspensions of titanium(IV) oxide nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text