Thermo Scientific Chemicals

Phenol, 99.5%, unstab., Thermo Scientific Chemicals

Catalog number: 044526.22
100 g, Each
Thermo Scientific Chemicals

Phenol, 99.5%, unstab., Thermo Scientific Chemicals

Catalog number: 044526.22
100 g, Each
Quantity
Catalog number: 044526.22
also known as 044526-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-95-2
IUPAC Name
phenol
Molecular Formula
C6H6O
InChI Key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1
Trace Metal
Heavy metals ( as Pb) 5 ppm, max
Assay (GC)
99.5%, min
Water Content (Karl Fischer Titration)
0.15%, max

Description

As a precursor to many materials and useful compounds. Its major uses involve its conversion to plastics or related materials. Phenol and its chemical derivatives are key for building polycarbonates, epoxies, Bakelite, nylon, detergents, and numerous pharmaceutical drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text