Thermo Scientific Chemicals

Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals

Catalog number: 044529.K2
1 L, Each
Thermo Scientific Chemicals

Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals

Catalog number: 044529.K2
1 L, Each
Quantity
Catalog number: 044529.K2
also known as 044529-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-21-1
IUPAC Name
ethane-1,2-diol
Molecular Formula
C2H6O2
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
OCCO
Optical Absorbance at
210 nm max, 1 AU
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
≥99%
Optical Absorbance at
220 nm max, 0.8 AU

Description

For use in spectrophotometryEthylene glycol is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text