Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals
Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
Catalog number 044529.M1
also known as 044529-M1
Price (USD)/ Each
238.65
Online exclusive
265.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
238.65
Online exclusive
265.00 
Save 26.35 (10%)
Add to cart
Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals
Catalog number044529.M1
Price (USD)/ Each
238.65
Online exclusive
265.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS107-21-1
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at210 nm max, 1 AU
Appearance (Color)Colorless
FormLiquid
Assay (GC)≥99%
Optical Absorbance at220 nm max, 0.8 AU
View more
For use in spectrophotometryEthylene glycol is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only