Thermo Scientific Chemicals

Zinc oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044533.09
10 g, Each
Thermo Scientific Chemicals

Zinc oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044533.09
10 g, Each
Quantity
Catalog number: 044533.09
also known as 044533-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-13-2
IUPAC Name
oxozinc
Molecular Formula
OZn
InChI Key
XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES
O=[Zn]
Total Metal Impurities
1% max.

Description

Zinc oxide is incorporated in powders, ointments, and pastes. As Accelerator activator, pigment and reinforcing agent in rubber, ointments, pigment and mould-growth inhibitor in paints, uv absorber in plastics, ceramics, floor tile, glass, zinc salts, feed additive, dietary supplement, seed treatment cosmetics, photoconductor in office copying machines and in color photography, piezoelectric devices, artists' colorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text