Thermo Scientific Chemicals

Polystyrene, atactic, Thermo Scientific Chemicals

Catalog number: 044537.09
10 g, Each
Thermo Scientific Chemicals

Polystyrene, atactic, Thermo Scientific Chemicals

Catalog number: 044537.09
10 g, Each
Quantity
Catalog number: 044537.09
also known as 044537-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-53-6
Molecular Formula
(C8H8)n
SMILES
*-CC(-*)C1=CC=CC=C1
MDL Number
MFCD00084450
Form
Beads
Infrared spectrum
Conforms
Molecular Weight
125000-250000

Description

Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text