Thermo Scientific Chemicals

Aluminum 2,4-pentanedionate, 99.995+% (metals basis), Thermo Scientific Chemicals

Catalog number: 044538.06
5 g, Each
Thermo Scientific Chemicals

Aluminum 2,4-pentanedionate, 99.995+% (metals basis), Thermo Scientific Chemicals

Catalog number: 044538.06
5 g, Each
Quantity
Catalog number: 044538.06
also known as 044538-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13963-57-0
IUPAC Name
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21AlO6
InChI Key
KILURZWTCGSYRE-LNTINUHCSA-K
SMILES
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Total Metal Impurities
≤0.005%
Appearance (Color)
White to pale yellow-green
Assay from Suppliers CofA
Report metals analysis
Form
Powder or crystals or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text