Thermo Scientific Chemicals
ICP-MS Water Spike Standard solution for 6020 CLP-M, Specpure™, Thermo Scientific Chemicals
Catalog number: 044539.AE
100 mL, Each
Thermo Scientific Chemicals
ICP-MS Water Spike Standard solution for 6020 CLP-M, Specpure™, Thermo Scientific Chemicals
Catalog number: 044539.AE
100 mL, Each
Quantity
Catalog number: 044539.AE
also known as 044539-AE
Price (USD)
Have Questions?
Concentration
Ba 100?g/mL ?2%
Concentration
Zn 10?g/mL ?2%
Concentration
Al 300?g/mL ?2%
Concentration
Be 10?g/mL ?2%
Concentration
Cd 10?g/mL ?2%