Thermo Scientific Chemicals

Retinoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 044540.02
0.5 g, Each
Thermo Scientific Chemicals

Retinoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 044540.02
0.5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 044540.02
also known as 044540-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
302-79-4
IUPAC Name
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular Formula
C20H28O2
InChI Key
SHGAZHPCJJPHSC-YCNIQYBTSA-N
SMILES
C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O
Mass Spec
Conforms to structure
Appearance (Color)
Pale yellow to yellow
Assay from Supplier's CofA
≥96.0%
Form
Powder or crystalline powder

Description

Retinoic acid is used in dermatology drugs. It is used in the treatment of psoriasis, ichthyosis, follicular keratosis, acne, lichen planus, verrucous epidermal nevus and lichen psoriasis. It is also employed in promoting the proliferation and differentiation of epithelial cells. In addition to this, it exhibits some preventive effect on skin cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient

• All-trans-retinol is involved in hormonal signaling via the retinoid receptors

• It is an important regulator of cell division and apoptosis

• Deficiency can lead to vision, bone, immune, and skin disorders

Applications

• Suitable for research on cell differentiation, embryonic development, immunology and more

• Laboratory grade quality, for research use only

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text