Retinoic acid, 97%, Thermo Scientific Chemicals
Retinoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Retinoic acid, 97%, Thermo Scientific Chemicals

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.1 g
0.5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044540.04
also known as 044540-04
Price (USD)/ Each
265.65
Online exclusive
295.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
265.65
Online exclusive
295.00 
Save 29.35 (10%)
Add to cart
Retinoic acid, 97%, Thermo Scientific Chemicals
Catalog number044540.04
Price (USD)/ Each
265.65
Online exclusive
295.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS302-79-4
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular FormulaC20H28O2
InChI KeySHGAZHPCJJPHSC-YCNIQYBTSA-N
SMILESC\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Mass SpecConforms to structure
Appearance (Color)Pale yellow to yellow
Assay from Supplier's CofA≥96.0%
FormPowder or crystalline powder
Retinoic acid is used in dermatology drugs. It is used in the treatment of psoriasis, ichthyosis, follicular keratosis, acne, lichen planus, verrucous epidermal nevus and lichen psoriasis. It is also employed in promoting the proliferation and differentiation of epithelial cells. In addition to this, it exhibits some preventive effect on skin cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient

• All-trans-retinol is involved in hormonal signaling via the retinoid receptors

• It is an important regulator of cell division and apoptosis

• Deficiency can lead to vision, bone, immune, and skin disorders

Applications

• Suitable for research on cell differentiation, embryonic development, immunology and more

• Laboratory grade quality, for research use only

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Stephensen, C.B.; Lietz, G. Vitamin A in resistance to and recovery from infection: relevance to SARS-CoV2. Br J Nutr. 2021, 126(11), 1663-1672.
  2. He, J.; Gu, Y.; Zhang, S. Vitamin A and Breast Cancer Survival: A Systematic Review and Meta-analysis. Clin Breast Cancer. 2018, 18(6), e1389-e1400.