Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 044556.36
Price (USD)/ Each
528.65
Online exclusive
587.00 
Save 58.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
528.65
Online exclusive
587.00 
Save 58.35 (10%)
Add to cart
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number044556.36
Price (USD)/ Each
528.65
Online exclusive
587.00 
Save 58.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only