4-Methyldiphenylamine, 98%, Thermo Scientific Chemicals
4-Methyldiphenylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyldiphenylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 044560.18
also known as 044560-18
Price (USD)/ Each
252.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
252.00
Add to cart
4-Methyldiphenylamine, 98%, Thermo Scientific Chemicals
Catalog number044560.18
Price (USD)/ Each
252.00
-
Add to cart
Specifications
Chemical Name or Material4-Methyldiphenylamine
CAS620-84-8
Health Hazard 1H302+H312-H315-H319
Health Hazard 2GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-Methyldiphenylamine is used as a syntheses material intermediates as well as OLED materials. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only