Thermo Scientific Chemicals

Aluminum sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044563.14
25 g, Each
Thermo Scientific Chemicals

Aluminum sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044563.14
25 g, Each
Quantity
Catalog number: 044563.14
also known as 044563-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-01-3
IUPAC Name
aluminium(3+) potassium disulfate
Molecular Formula
AlKO8S2
InChI Key
GRLPQNLYRHEGIJ-UHFFFAOYSA-J
SMILES
[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Metal Impurities
≤0.01% (100 ppm)
Appearance (Color)
White
Form
Powder or chunks
Assay (special)
?2.5 % of Theoretical value (Al)

Description

Aluminum sulfate acts as a flocculating agent in the purification of drinking water, and in waste water treatment plants. It acts as a mordant in dyeing, printing, textiles and also used in paper manufacturing. It is a waterproofing agent and accelerator in concrete. Further, it is used as a foaming agent in fire fighting foam, photographic film and in photochemicals. It is also used in styptic pencils, pain relief and in dentistry for gingival retraction cords.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text