Thermo Scientific Chemicals

Polyphenylmethylsiloxane, MW 2500-2700, Thermo Scientific Chemicals

Catalog number: 044569.30
250 g, Each
Thermo Scientific Chemicals

Polyphenylmethylsiloxane, MW 2500-2700, Thermo Scientific Chemicals

Catalog number: 044569.30
250 g, Each
Quantity
Catalog number: 044569.30
also known as 044569-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-12-3
IUPAC Name
methyl(phenyl)silanone
Molecular Formula
C7H8OSi
InChI Key
IXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILES
C[Si](=O)C1=CC=CC=C1
Appearance (Color)
Clear, colorless
Form
Viscous liquid
Viscosity
450-550 cSt
Infrared spectrum
Conforms to structure

Description

A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text