Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals
Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044575.14
also known as 044575-14
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
Add to cart
Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals
Catalog number044575.14
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPellets
Total Metal Impurities≤0.01%
Appearance (Color)Grey to black
Assay from Supplier's CofA1.90-2.10% (ruthemium, gravimetric)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only