Chromium etchant, Thermo Scientific Chemicals
Chromium etchant, Thermo Scientific Chemicals
Thermo Scientific™

Chromium etchant, Thermo Scientific Chemicals

High Purity Ceric Ammonium Nitrate System For Precise, Clean Etching of Chromium and Chromium Oxide Films
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 qt.
1 gal.
Catalog number 044582.R0
also known as 044582-R0
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 qt.
Request bulk or custom format
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
Add to cart
Chromium etchant, Thermo Scientific Chemicals
Catalog number044582.R0
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialChromium etchant
ColorOrange
Health Hazard 1H272-H314: May intensify fire; oxidizer. Causes severe skin burns and eye damage.
DOT InformationDOT Class: 8, Packaging Group: II
MDL NumberMFCD07370678
View more
High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only