Copper etchant, Thermo Scientific Chemicals
Copper etchant, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper etchant, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 gal.
1 qt.
Catalog number 044583.LV
also known as 044583-LV
Price (USD)/ Each
213.65
Online exclusive
237.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
1 gal.
Request bulk or custom format
Price (USD)/ Each
213.65
Online exclusive
237.00 
Save 23.35 (10%)
Add to cart
Copper etchant, Thermo Scientific Chemicals
Catalog number044583.LV
Price (USD)/ Each
213.65
Online exclusive
237.00 
Save 23.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCopper etchant
Health Hazard 1H290-H302-H315-H317-H318
Health Hazard 3P234-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P390-P501c
Recommended StorageAmbient temperatures
DOT InformationHazard Class: 8; Packaging Group: III
View more
Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only