Gold etchant, Thermo Scientific Chemicals
Gold etchant, Thermo Scientific Chemicals
Thermo Scientific™

Gold etchant, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 gal.
1 qt.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044584.LV
also known as 044584-LV
Price (USD)/ Each
748.00
-
Add to cart
Quantity:
1 gal.
Request bulk or custom format
Price (USD)/ Each
748.00
Add to cart
Gold etchant, Thermo Scientific Chemicals
Catalog number044584.LV
Price (USD)/ Each
748.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGold etchant
MDL NumberMFCD07370792
Quantity1 gal.
FormLiquid
Unit SizeEach
High purity, low sodium and 0.2 micron filtration system for semiconductor and microelectronic applicationsGold etchant is a selective general-purpose etchant for gold. It is compatible with negative and positive photoresists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only