Nichrome etchant, Thermo Scientific Chemicals
Nichrome etchant, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nichrome etchant, Thermo Scientific Chemicals

High Purity Ceric Ammonium Nitrate System Provides Clean, Precise Etching of Nichrome Layers For Preparation of Thin Film Circuits
Have Questions?
Quantity:
1 gal.
Catalog number 044585.LV
also known as 044585-LV
Price (USD)/ Each
403.65
Online exclusive
448.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
1 gal.
Request bulk or custom format
Price (USD)/ Each
403.65
Online exclusive
448.00 
Save 44.35 (10%)
Add to cart
Nichrome etchant, Thermo Scientific Chemicals
Catalog number044585.LV
Price (USD)/ Each
403.65
Online exclusive
448.00 
Save 44.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialNichrome etchant
ColorColorless
Health Hazard 1H290-H314-H331-H360D-H362
Health Hazard 2When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Health Hazard 3P201-P202-P234-P260-P263-P264b-P270-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
High purity ceric ammonium nitrate system provides clean, precise etching of nichrome layers for preparation of thin film circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only