Nickel etchant for evaporated nickel films, Thermo Scientific Chemicals
Nickel etchant for evaporated nickel films, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel etchant for evaporated nickel films, Thermo Scientific Chemicals

Standard nickel etchant for evaporated nickel films
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 qt.
1 gal.
Catalog number 044586.R0
also known as 044586-R0
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 qt.
Request bulk or custom format
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
Add to cart
Nickel etchant for evaporated nickel films, Thermo Scientific Chemicals
Catalog number044586.R0
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialNickel etchant for evaporated nickel films
ColorColorless to Yellow
Health Hazard 1H290-H314-H331-H360D-H362
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P201-P202-P234-P260-P263-P264b-P270-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Standard nickel etchant for evaporated nickel films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only