Thermo Scientific Chemicals

Neodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 044587.18
50 g, Each
Thermo Scientific Chemicals

Neodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 044587.18
50 g, Each
Quantity
Catalog number: 044587.18
also known as 044587-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
334869-71-5
IUPAC Name
neodymium(3+) triacetate
Molecular Formula
C6H9NdO6
InChI Key
KETUDCKOKOGBJB-UHFFFAOYSA-K
SMILES
[Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text