Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 044588.30
also known as 044588-30
Price (USD)/ Each
1,693.65
Online exclusive
1882.00 
Save 188.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
1,693.65
Online exclusive
1882.00 
Save 188.35 (10%)
Add to cart
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Catalog number044588.30
Price (USD)/ Each
1,693.65
Online exclusive
1882.00 
Save 188.35 (10%)
-
Add to cart
Chemical Identifiers
CAS11140-77-5
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.