Thermo Scientific Chemicals

Yttrium(III) trifluoroacetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 044597.09
10 g, Each
Thermo Scientific Chemicals

Yttrium(III) trifluoroacetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 044597.09
10 g, Each
Quantity
Catalog number: 044597.09
also known as 044597-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
304851-95-4
IUPAC Name
yttrium(3+) tritrifluoroacetate
Molecular Formula
C6F9O6Y
InChI Key
NPATVYPXYMSPKD-UHFFFAOYSA-K
SMILES
[Y+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Total Rare Earth Oxide Impurities
0.1% max.

Description

A fluorinated yttrium compound for proteomics research. Used in the manufacture of ceramics, glass, laser materials, phosphor. non-ferrous alloy iron and steel, and the additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text