Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

For Synthesis and Liquid Chromatography
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
100 mL
1 L
Catalog number 044608.M1
also known as 044608-M1
Price (USD)/ Each
313.65
Online exclusive
348.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
313.65
Online exclusive
348.00 
Save 34.35 (10%)
Add to cart
Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number044608.M1
Price (USD)/ Each
313.65
Online exclusive
348.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
CommentInert gas used can be either Argon or Nitrogen
Water Content (Karl Fischer Titration)≤50 ppm
StabilizerBHT (report only)
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only