Thermo Scientific Chemicals

2-Amino-5-bromo-4-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 044623.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-5-bromo-4-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 044623.14
25 g, Each
Quantity
Catalog number: 044623.14
also known as 044623-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98198-48-2
IUPAC Name
2-amino-5-bromo-4-methylpyridin-1-ium
Molecular Formula
C6H8BrN2
InChI Key
JDNCMHOKWINDKI-UHFFFAOYSA-O
SMILES
CC1=CC(N)=[NH+]C=C1Br
Appearance (Color)
Very pale cream to light brown
Form
Powder
Assay (unspecified)
> 96.0%

Description

2-Amino-5-bromo-4-methylpyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text