Buffer HF-Improved, Thermo Scientific Chemicals
Buffer HF-Improved, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer HF-Improved, Thermo Scientific Chemicals

Used For Pretreatment of Planar Silicon Devices When Plating With Nickel Plating Solution Products 44069 and 44070
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 044627.AK
Price (USD)/ Each
-
Quantity:
250 mL
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialBuffer HF-Improved
Boiling Point103°C to 105°C
Health Hazard 1H290-H301+H331-H310-H314-H335
ColorColorless
Health Hazard 3P234-P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
View more
Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only